Skip to Content
Merck
All Photos(1)

Key Documents

07-748-AF647

Sigma-Aldrich

Anti-monomethyl Histone H4 (Lys20) Antibody, Alexa Fluor 647 Conjugate

0.5 mg/mL, from rabbit

Synonym(s):

Histone H4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 647

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

concentration

0.5 mg/mL

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

monomethylation (Lys20)

Gene Information

General description

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The histones have an amino terminal tail, a globular domain, and a carboxy-terminal tail. The four core histones, H2A, H2B, H3 and H4 assemble into the octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped in a sequence-independent manner around the octamer, forming the basic subunit of chromatin, the nucleosome. Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail, H4 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. This product is a conjugated version of Cat. No. 07-748.

Immunogen

Linear peptide corresponding to Histone H4 monomethylated at Lys20.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
This Anti-monomethyl Histone H4 (Lys20) Antibody, Alexa Fluor 647 Conjugate is validated for use in ICC for the detection of monomethyl Histone H4 (Lys20).

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected monomethyl Histone H4 (Lys20) in HeLa cells.

Alexa Fluor is a registered trademark of Life Technologies.

Target description

~10 kDa observed (Refer to Cat. No. 07-748)

Physical form

Protein A purified
Purified rabbit conjugated antibody in buffer containing PBS and 15 mg/ml BSA with 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service