Skip to Content
Merck
All Photos(4)

Key Documents

07-213-I

Sigma-Aldrich

Anti-dimethyl-Histone H4 (Arg3) Asymmetric Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

technique(s)

dot blot: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

dimethylation (Arg3)

Gene Information

General description

Histone H4 has multiple isoform names including H4/A through H4/N, and is encoded by multiple genes in the genome named HIST1H4A through HIST1H4N/H4/A or H4FA through H4/L and HIST2H4A, HIST2H4B, and HIST4H4. Histone H4 is a critical component of chromatin and the nucleosome. Histone H4 forms a multimeric cluster with the other histone members to regulate chromatin and DNA structure within the nucleus. Histone H4 is a target of much post-translational modification including acetylation, methylation, phosphorylation, ubiquitination, sumoylation, crotonylation (male germ cells only and marks testis specific genes) and citrullination. Acetylation of histone H4 on lysine 16 (H4-K16Ac) is the most prevalent and reversible posttranslational chromatin modification in eukaryotes for histone H4 and it and the other modifications help control chromatin structure and the protein interactions of histone H4.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of human dimethyl-Histone H4 (Arg3) Asymmetric.

Application

Dot Blot Specificity Analysis: Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665), which contain histone peptides with various modifications were probed with dimethyl-Histone H4 (Arg3) Asymmetric (1:2,000 dilution).
Immunoprecipitation Analysis: A 1:3,333 dilution from a representative lot immunoprecipitated dimethyl-Histone H4 (Arg3) Asymmetric in 500 µg of HeLa and 500ng of HeLa AE cell lysate.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
This Anti-dimethyl-Histone H4 (Arg3) Asymmetric Antibody is validated for use in Western Blotting and Immunoprecipitation and Dot Blot for the detection of dimethyl-Histone H4 (Arg3) Asymmetric.

Quality

Evaluated by Western Blotting of Histone H4 recombinant protein.

Western Blotting Analysis: A 1:3,333 dilution of this antibody detected dimethyl-Histone H4 (Arg3) Asymmetric in 500 ng of PRMT1 treated Histone H4 recombinant protein.

Target description

~13 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng-Han Yu et al.
Bioscience reports, 38(2) (2018-03-16)
Phenyl vinyl sulfone (PVS) and phenyl vinyl sulfonate (PVSN) inactivate protein tyrosine phosphatases (PTPs) by mimicking the phosphotyrosine structure and providing a Michael addition acceptor for the active-site cysteine residue of PTPs, thus forming covalent adducts between PVS (or PVSN)
Daniele Musiani et al.
Cell reports, 30(4), 1208-1222 (2020-01-30)
Protein arginine methyltransferase 1 (PRMT1) is overexpressed in various human cancers and linked to poor response to chemotherapy. Various PRMT1 inhibitors are currently under development; yet, we do not fully understand the mechanisms underpinning PRMT1 involvement in tumorigenesis and chemoresistance.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service