Skip to Content
Merck
All Photos(2)

Key Documents

06-966

Sigma-Aldrich

Anti-CDK1/CDC2 Antibody (C-Term)

serum, Upstate®

Synonym(s):

Cyclin-dependent kinase 1, CDK1, Cell division control protein 2 homolog, Cell division protein kinase 1, p34 protein kinase, Cell division control protein 2 homolog, Cell division protein kinase 1, p34 protein kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

species reactivity (predicted by homology)

hamster (with greater than 90% sequence homology), human (with greater than 90% sequence homology), rat (with greater than 90% sequence homology)

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

hamster ... Cdk1(100766716)
human ... CDK1(983)
mouse ... Cdk1(12534)
rat ... Cdk1(54237)

General description

Entry of all eukaryotic cells into mitosis is regulated by activation of CDC2 kinase. Activation of CDC2 is controlled at several steps including cyclin binding and phosphorylation of Thr161. However, the critical regulatory step in activating CDC2 during progression into mitosis appears to be dephosphorylation of Tyr15 and Thr14. Phosphorylation at Tyr15 and inhibition of CDC2 is carried out by Wee1 and Myt1 protein kinases, while Tyr15 dephosphorylation and activation of CDC2 is carried out by the CDC25 phosphatase. Expression of CDC2 plays a critical role in cell transition through the G2/M phase. The activity of CDC2 in dividing cells increases at the onset of M phase coincident with disassembly of the cell nucleus, generation of mitotic spindles and chromosome condensation. Several proteins including Histone H1 and pp60src are substrates for CDC2.

Immunogen

KLH-conjugated linear peptide corresponding to a region near the near the C-terminus of human CDK1/CDC2.

Application

Detect Cdk1/Cdc2 with Anti-Cdk1/Cdc2 Antibody, CT (Rabbit Polyclonal Antibody), that has been shown to work in IP & WB.
Immunoprecipitation Analysis: 5 µL of this antibody immunoprecipited CDK1/CDC2 (C-terminus) from Balb/3T3/A31 cell lysate. Immunoprecipited sample was then subject to Western Blotting analysis.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Western Blotting in Balb/3T3/A31 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CDK1/CDC2 (C-terminus) in Balb/3T3/A31 cell lysate.

Target description

34kDa

Physical form

Antiserum
rabbit antiserum containing 0.05% sodium azide

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Positive Antigen Control: Catalog #12-305, 3T3/A31 lysate. Add 2.5 μL of 2-mercapto-ethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mismatch repair status and the response of human cells to cisplatin.
Elisabetta Pani,Lovorka Stojic,Mahmoud El-Shemerly,Josef Jiricny,Stefano Ferrari
Cell Cycle null
RB activation defect in tumor cell lines.
Broce?o, C; Wilkie, S; Mittnacht, S
Proceedings of the National Academy of Sciences of the USA null
Isabelle Demeestere et al.
Biology of reproduction, 87(1), 3-3 (2012-04-21)
During folliculogenesis, oocytes grow and acquire developmental competence in a mutually dependent relationship with their adjacent somatic cells. Follicle-stimulating hormone (FSH) plays an essential and well-established role in the differentiation of somatic follicular cells, but its function in the development
Proteasomal activity is required to initiate and to sustain translational activation of messenger RNA encoding the stem-loop-binding protein during meiotic maturation in mice.
Yang, Q; Allard, P; Huang, M; Zhang, W; Clarke, HJ
Biology of Reproduction null
Stephanie Wallace et al.
ACS sensors, 8(9), 3338-3348 (2023-08-23)
Our growing ability to tailor healthcare to the needs of individuals has the potential to transform clinical treatment. However, the measurement of multiple biomarkers to inform clinical decisions requires rapid, effective, and affordable diagnostics. Chronic diseases and rapidly evolving pathogens

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service