Skip to Content
Merck
All Photos(4)

Documents

06-725-I

Sigma-Aldrich

Anti-FcεRI α subunit Antibody

serum, from rabbit

Synonym(s):

High affinity immunoglobulin epsilon receptor subunit alpha, Fc-epsilon RI-alpha, FcERI, IgE Fc receptor subunit alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FCER1A(2205)

General description

The allergic reaction is mediated through the IgE high affinity receptor (FcεRI). When a multivalent antigen is presented, a redistribution of the monomeric IgE FcεRI receptor complexes causes the degranulation of mast cells and basophils, resulting in the subsequent release of factors responsible for the allergic reaction. The FcεRI receptor is a tetrameric complex, consisting of an a-chain, b-chain and a dimeric g-chain. The a-chain is primarily involved with the binding of IgE, but the signal is transferred through the g subunit, which contains the immunoreceptor tyrosine activation motifs (ITAM) critical for initiating receptor mediated signal transduction through the Syk and Lyn tyrosine kinases. The FcεRI receptor is able to engage and disengage the IgE, providing a versatile model for studying the function of kinase coupled receptors.

Specificity

This antibody recognizes the extracellular domain of FcεRI α subunit.

Immunogen

Epitope: Extracellular domain
GST-tagged recombinant protein corresponding to the extracellular domain of human FcεRI α subunit.

Application

Detect FcεRI α subunit using this Anti-FcεRI α subunit Antibody validated for use in Western Blotting, Immunoprecipitation, ICC.
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms
Western Blot Analysis: A 1:5,000 dilution from a representative lot detected FcεRI α subunit in 10 µg of human spleen tissue lysate.

Immunoprecipitation Analysis: A 1:50 dilution from a representative lot immunoprecipitated FcεRI α subunit from human spleen whole lysate.

Flow Cytometry Analysis: A 1:200 dilution from a representative lot detected FcεRI α subunit in RBL cells.

Quality

Evaluated by Western Blot in RBL cell lysate.

Western Blot Analysis: A 1:5,000 dilution of this antibody detected FcεRI α subunit in 10 µg of RBL cell lysate.

Target description

~57 kDa observed

Linkage

Replaces: 06-725

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
RBL cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service