Skip to Content
Merck
All Photos(1)

Key Documents

05-901R

Sigma-Aldrich

Anti-NR2A Antibody, clone A12W, rabbit monoclonal

Synonym(s):

N-methyl D-aspartate receptor subtype 2A, N-methyl-D-aspartate receptor channel, subunit epsilon-1, N-methyl-D-aspartate receptor subunit 2A, NMDA receptor subtype 2A, glutamate receptor, ionotropic, N-methyl D-aspartate 2A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

antibody product type

primary antibodies

form

liquid

target post-translational modification

unmodified

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Chen et al.
Journal of neuroinflammation, 19(1), 181-181 (2022-07-14)
Perioperative neurocognitive disorders (PNDs) are considered the most common postoperative complication in geriatric patients. However, its pathogenesis is not fully understood. Surgery-triggered neuroinflammation is a major contributor to the development of PNDs. Neuroinflammation can influence N-methyl-D-aspartate receptor (NMDAR) expression or
Han Zhao et al.
Brain and behavior, 8(7), e01004-e01004 (2018-06-02)
It is known that an interruption of acoustic input in early life will result in abnormal development of the auditory system. Here, we further show that this negative impact actually spans beyond the auditory system to the hippocampus, a system
Li-Hsin Chang et al.
Scientific reports, 8(1), 118-118 (2018-01-10)
Inhibition and deletion of soluble epoxide hydrolase (sEH) has been suggested to ameliorate infarction in experimental ischemic stroke possibly via vasoactive epoxyeicosatrienoic acids. However, it is unknown whether the neuroprotective mechanisms involve alteration of post-ischemic neuronal transmission and neurotrophic signaling.
Yukio Hattori et al.
The journal of obstetrics and gynaecology research, 43(2), 412-415 (2017-02-06)
Anti-N-methyl-d-aspartate receptor (NMDAR) limbic encephalitis is the most common form of paraneoplastic encephalitis that is associated with teratomas. Because tumor removal leads to better clinical outcomes, it is essential to reveal the location of the teratomas. This is the first
Teri M Furlong et al.
Addiction biology, 23(3), 857-867 (2017-07-15)
Goal-directed actions are controlled by the value of the consequences they produce and so increase when what they produce is valuable and decrease when it is not. With continued invariant practice, however, goal-directed actions can become habits, controlled not by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service