Skip to Content
Merck
All Photos(3)

Documents

04-638

Sigma-Aldrich

Anti-TRF1 Antibody, clone BED5 57-6

clone BED5 57-6, Upstate®, from mouse

Synonym(s):

TERF1, telomeric repeat binding factor (NIMA-interacting) 1, FLJ41416, PIN2, TRBF1, TRF, TRF1, hTRF1-AS, t-TRF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

BED5 57-6, monoclonal

species reactivity

mouse, rat, hamster, monkey, human

packaging

antibody small pack of 25 μg

manufacturer/tradename

Upstate®

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TERF1(7013)

Specificity

Detects TRF1

Immunogen

Human TRF1 protein

Application

This Anti-TRF1 Antibody, clone BED5 57-6 is validated for use in WB, IF for the detection of TRF1.

Quality

Routinely evaluated by Western Blot on HeLa cell lysate.

Target description

~60 kDa

Physical form

Affinity purified immunoglobulin in PBS with 0.05% NaN3
Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PTOP and TRF1 help enhance the radio resistance in breast cancer cell.
Li, Z; Yang, X; Xia, N; Yang, L; Yu, H; Zhou, F; X, C; Zhou, Y
Cancer Cell International null
Emily Mason-Osann et al.
EMBO reports, 21(6), e49495-e49495 (2020-04-28)
Cancer cells can activate the alternative lengthening of telomeres (ALT) pathway to promote replicative immortality. The ALT pathway promotes telomere elongation through a homologous recombination pathway known as break-induced replication (BIR), which is often engaged to repair single-ended double-stranded breaks

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service