Skip to Content
Merck
All Photos(3)

Key Documents

04-1643

Sigma-Aldrich

Rabbit Anti-Mouse IgG Antibody, clone RM104

clone RM104, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

secondary antibodies

clone

RM104, monoclonal

species reactivity

mouse

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Specificity

This antibody reacts to the Fc region of all subclasses of mouse IgG. No cross reactivity with mouse IgM, IgA, IgE, human IgG, and rat IgG. It may cross react to goat IgG. The Fc region of RM104 has been engineered to eliminate the binding of Fc receptor.

Immunogen

Mouse IgG

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling
This rabbit monoclonal Rabbit Anti-Mouse IgG, clone RM104, Cat. No. 04-1643 is validated for use in ELISA and Western Blotting, for the detection of Mouse IgG.
Western Blotting Analysis (WB): 0.2 μg/mL from a representative lot detected Mouse IgG in nonreduced (-) mouse immunoglobulins.

Quality

ELISA Analysis: The specificity and reactivity of a representative lot was confirmed by ELISA analyses.

Physical form

Format: Purified
Protein A purified
Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service