JWPH3249DD75
J & W Pharmlab LLC
1-(3-Methoxy-pyridin-2-yl)-piperazine dihydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1-(3-methoxy-2-pyridinyl)piperazine dihydrochloride
Inchi Code
1S/C10H15N3O.2ClH/c1-14-9-3-2-4-12-10(9)13-7-5-11-6-8-13;;/h2-4,11H,5-8H2,1H3;2*1H
InChI key
XUIKTDZQHPBVKN-UHFFFAOYSA-N
Purity
96%
Country of Origin
US
Physical Form
off white solid
Product Link
https://www.jwpharmlab.com/home/product/65R0038S
MSDS
https://www.jwpharmlab.com/home/product/65R0038S
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service