Skip to Content
Merck
All Photos(1)

Documents

JWPH3249C19D

J & W Pharmlab LLC

3-Bromo-2-bromomethyl-4-methoxy-benzoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
338
MDL number:

Iupac Name

methyl 3-bromo-2-(bromomethyl)-4-methoxybenzoate

Inchi Code

1S/C10H10Br2O3/c1-14-8-4-3-6(10(13)15-2)7(5-11)9(8)12/h3-4H,5H2,1-2H3

InChI key

XAABTWWFSLPRFK-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

White Solid

Product Link

https://www.jwpharmlab.com/home/product/03R0378

MSDS

https://www.jwpharmlab.com/home/product/03R0378


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service