Skip to Content
Merck
All Photos(1)

Documents

CIAH998D8195

ChemScene LLC (Preferred Partner)

Diethyl 2-formylsuccinate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
202.21
MDL number:

bp

260.32 C at 760 mmHg

Iupac Name

diethyl (2E)-2-(hydroxymethylene)butanedioate

Inchi Code

1S/C9H14O5/c1-3-13-8(11)5-7(6-10)9(12)14-4-2/h6-7H,3-5H2,1-2H3

InChI key

LHHLYOVYBZWIGM-UHFFFAOYSA-N

storage temp.

-20C, stored under nitrogen

Salt Data

Succinate

Purity

96%

Country of Origin

CN

Physical Form

Liquid

Product Link

https://www.chemscene.com/5472-38-8.html

Boiling point

260.32 C at 760 mmHg

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0140696.pdf


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service