Skip to Content
Merck
All Photos(1)

Documents

CH6447918588

Chem Impex International, Inc.

Maleimido-3,5-disuccinimidyl isophthalate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
455.34
MDL number:

mp

247 C (Decomp)

Iupac Name

1-(3,5-bis{[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl}phenyl)-1H-pyrrole-2,5-dione

Inchi Code

1S/C20H13N3O10/c24-13-1-2-14(25)21(13)12-8-10(19(30)32-22-15(26)3-4-16(22)27)7-11(9-12)20(31)33-23-17(28)5-6-18(23)29/h1-2,7-9H,3-6H2

InChI key

SZHLOZJGLNEUBJ-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

95%

Shipping Temperature

RT

Physical Form

Off-white solid

Product Link

https://www.chemimpex.com/product/productinfo/maleimido-3-5-disuccinimidyl-isophthalate/13655?cid=0

Melting Point

247 C (Decomp)

MSDS

https://www.chemimpex.com/product/productinfo/maleimido-3-5-disuccinimidyl-isophthalate/13655?cid=0


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service