Skip to Content
Merck
All Photos(1)

Key Documents

Z-019

Supelco

Ziprasidone-D8 hydrochloride solution

100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Labeled Ziprasidone HCl

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
R$1,333.00

R$1,333.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
R$1,333.00

About This Item

Empirical Formula (Hill Notation):
C21H13D8ClN4OS · HCl
Molecular Weight:
457.45
UNSPSC Code:
41116107
NACRES:
NA.24

R$1,333.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

General description

Ziprasidone is sold under the trade names Geodon and Zeldox for treatment of schizophrenia and bipolar disorder. Recent studies have suggested that Ziprasidone may increase the risk of Type II diabetes and hyperglycemia. A certified Snap-N-Shoot® solution suitable for quantification of ziprasidone by LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, or forensic analysis applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service