Skip to Content
Merck
All Photos(1)

Documents

V-068

Supelco

Valeryl fentanyl-D5 hydrochloride solution

100 μg/mL in methanol (as free base), certified reference material, ampule of 0.5 mL, Cerilliant®

Synonym(s):

Deuterated Valeryl Fentanyl HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27D5N2O · HCl
Molecular Weight:
406.02
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 0.5 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

General description

Valeryl fentanyl is a designer drug and fentanyl analog. In March 2017, Chinese officials banned manufacturing and sale of valeryl fentanyl in China. This stable-labeled internal standard is suitable for quantification of valeryl fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, or forensic analysis applications. This Certified Spiking Solution; is supplied in a convenient US DEA-exempt solution format.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service