Skip to Content
Merck
All Photos(1)

Documents

T-060

Supelco

Tapentadol-β-D-glucuronide solution

100 μg/mL in acetonitrile: water, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H31NO7
CAS Number:
Molecular Weight:
397.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in acetonitrile: water

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CC[C@H]([C@@H](C)CN(C)C)c1cccc(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(O)=O)c1

InChI

1S/C20H31NO7/c1-5-14(11(2)10-21(3)4)12-7-6-8-13(9-12)27-20-17(24)15(22)16(23)18(28-20)19(25)26/h6-9,11,14-18,20,22-24H,5,10H2,1-4H3,(H,25,26)/t11-,14+,15-,16-,17+,18-,20+/m0/s1

InChI key

CTYJDHSTNLOUMT-PJOQTPGLSA-N

General description

The first Certified Spiking Solution® of the primary urinary metabolite of tapentadol. Tapentadol is the first new drug of the centrally-acting analgesic class approved in the US for pain management in more than 25 years. This Certified Spiking Solution® is suitable for many LC/MS or GC/MS applications, including pain prescription monitoring, forensic testing, clinical toxicology, or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service