Skip to Content
Merck
All Photos(1)

Key Documents

S-079

Supelco

UR-144 5-Hydroxypentyl metabolite-D5 (indole-D5) solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

UR-144 Metabolite Solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
R$1,208.00

R$1,208.00


Estimated to ship onApril 24, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
R$1,208.00

About This Item

Empirical Formula (Hill Notation):
C21H24D5NO2
Molecular Weight:
332.49
UNSPSC Code:
41116107
NACRES:
NA.24

R$1,208.00


Estimated to ship onApril 24, 2025


Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

General description

An internal standard suitable for quantitation of UR-144 5-Hydroxypentyl metabolite levels using LC-MS/MS or GC/MS for synthetic cannabinoid testing methods in forensic analysis, clinical toxicology, or urine drug testing. UR-144 5-Hydroxypentyl metabolite is a primary urinary metabolite of UR-144, a third generation synthetic cannabinoid that has seen wide popularity in illicit drug markets since late 2012. This biomarker is also referred to as the terminal omega hydroxyl metabolite of UR-144.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service