Skip to Content
Merck
All Photos(1)

Key Documents

P-009

Supelco

Pentobarbital-D5 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
R$266.00

R$266.00


Estimated to ship onApril 11, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
R$266.00

About This Item

Empirical Formula (Hill Notation):
C11D5H13N2O3
CAS Number:
Molecular Weight:
231.30
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

R$266.00


Estimated to ship onApril 11, 2025


Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C1(C(C)CCC)C(=O)NC(=O)NC1=O

InChI

1S/C11H18N2O3/c1-4-6-7(3)11(5-2)8(14)12-10(16)13-9(11)15/h7H,4-6H2,1-3H3,(H2,12,13,14,15,16)/i2D3,5D2

InChI key

WEXRUCMBJFQVBZ-ZTIZGVCASA-N

Looking for similar products? Visit Product Comparison Guide

General description

A stable-labeled internal standard suitable for pentobarbital testing by GC/MS or LC/MS for clinical toxicology, forensic analysis, or urine drug testing applications. Pentobarbital is a short-acting barbiturate and hypnotic sold under the trade name Nembutal® and approved for treatment of seizures and preoperative sedation. Also known as pentobarbitone, the sodium salt of pentobarbital is used as an anesthetic in veterinary medicine.

Application


  • Pentobarbital-D₅ as a reference standard in therapeutic drug monitoring: Pentobarbital-D₅ is utilized as a stable isotope-labeled internal standard in the quantitative determination of drugs, including statins, by high-resolution tandem mass spectrometry. This application is critical for adherence testing and therapeutic drug monitoring, ensuring precise and reliable measurement in biochemical research (Wagmann et al., 2020).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Nembutal is a registered trademark of Oak Pharmaceuticals, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service