Skip to Content
Merck
All Photos(1)

Key Documents

N-922

Supelco

Norfluoxetine-D6 oxalate solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Labeled Norfluoxetine Oxalate

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
R$329.00

R$329.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
R$329.00

About This Item

Empirical Formula (Hill Notation):
C16D6H10F3NO · C2H2O4
Molecular Weight:
391.37
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

R$329.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

NCCC(C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])([2H])OC2=CC=C(C(F)(F)F)C=C2.OC(C(O)=O)=O

InChI

1S/C16H16F3NO.C2H2O4/c17-16(18,19)13-6-8-14(9-7-13)21-15(10-11-20)12-4-2-1-3-5-12;3-1(4)2(5)6/h1-9,15H,10-11,20H2;(H,3,4)(H,5,6)/i1D,2D,3D,4D,5D,15D;

InChI key

FDWJCEPFCGIDPF-OYCZTGALSA-N

General description

Norfluoxetine is a primary blood metabolite of the selective serotonin reuptake inhibitor (SSRI) fluoxetine, an antidepressant, sold under the trade name Prozac®. This internal standard is suitable for quantitation of fluoxetine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Prozac is a registered trademark of Eli Lilly and Co.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service