Skip to Content
Merck
All Photos(1)

Key Documents

N-078

Supelco

(S)-N-Nitrosoanatabine solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

NAT

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
R$1,580.00

R$1,580.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
R$1,580.00

About This Item

Empirical Formula (Hill Notation):
C10H11N3O
CAS Number:
Molecular Weight:
189.21
UNSPSC Code:
41116107
NACRES:
NA.24

R$1,580.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

N1([C@@H](CC=CC1)c2cnccc2)N=O

InChI

1S/C10H11N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h1-4,6,8,10H,5,7H2/t10-/m0/s1

InChI key

ZJOFAFWTOKDIFH-JTQLQIEISA-N

General description

N-Nitrosoanatabine or NAT is a potent carcinogen and biomarker of exposure to cigarette smoke. It′s found in a variety of tobacco products such as cigarettes, electronic cigarettes, nicotine gum and oral snuff. This certified Snap-N-Spike® solution is suitable for use in LC-MS/MS or GC/MS methods for nicotine testing, environmental monitoring, or clinical and diagnostic testing of nicotine biomarkers.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service