Skip to Content
Merck
All Photos(1)

Key Documents

L-051

Supelco

Lormetazepam-13C,D3 solution

100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
R$1,904.00

R$1,904.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
R$1,904.00

About This Item

Empirical Formula (Hill Notation):
C1513CH9D3Cl2N2O2
CAS Number:
Molecular Weight:
339.20
UNSPSC Code:
41116107
NACRES:
NA.24

R$1,904.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=CC(C(C2=CC=CC=C2Cl)=NC(O)C3=O)=C(C=C1)N3[13C]([2H])([2H])[2H]

InChI

1S/C16H12Cl2N2O2/c1-20-13-7-6-9(17)8-11(13)14(19-15(21)16(20)22)10-4-2-3-5-12(10)18/h2-8,15,21H,1H3/i1+1D3

InChI key

FJIKWRGCXUCUIG-KQORAOOSSA-N

General description

Lormetazepam is a benzodiazepine with sedative-hypnotic, anxiolytic, anticonvulsant, and muscle relaxant effects. Lormetazepam is sold under trade names such as Loramet and Noctamid for treatment of insomnia. This Snap-N-Shoot® stable-labeled internal standard is suitable for quantitation of lormetazepam levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service