Skip to Content
Merck
All Photos(1)

Documents

ERA-013K

Supelco

Aldehyde/Ketone-DNPH Stock Standard-13

3 μg/mL each component in acetonitrile (as aldehyde/ketone), ampule of 5 × 1.2 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107

grade

certified reference material

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

Acetaldehyde-DNPH
Acetone-DNPH
Acrolein-DNPH
Benzaldehyde-DNPH
2-Butanone-DNPH
n-Butyraldehyde-DNPH
Crotonaldehyde-DNPH
Formaldehyde-DNPH
Hexaldehyde-DNPH
Methacrolein-DNPH
Propionaldehyde-DNPH
m-Tolualdehyde-DNPH
Valeraldehyde-DNPH

packaging

ampule of 5 × 1.2 mL

manufacturer/tradename

Cerilliant®

concentration

3 μg/mL each component in acetonitrile (as aldehyde/ketone)

application(s)

environmental

format

multi-component solution

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    2-Butanone-2,4-dinitrophenylhydrazone
    Acetaldehyde-2,4-dinitrophenylhydrazone
    Acetone-2,4-dinitrophenylhydrazone
    Acrolein-2,4-dinitrophenylhydrazone
    Benzaldehyde-2,4-dinitrophenylhydrazone
    Crotonaldehyde-2,4-dinitrophenylhydrazone
    Hexaldehyde-2,4-dinitrophenylhydrazone
    Methacrolein-2,4-dinitrophenylhydrazone
    m-Tolualdehyde 2,4-dinitrophenylhydrazone
    Butyraldehyde-2,4-dinitrophenylhydrazone
    Propionaldehyde-2,4-dinitrophenylhydrazone
    Valeraldehyde-2,4-dinitrophenylhydrazone
    See All (12)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service