Skip to Content
Merck
All Photos(1)

Key Documents

C-168

Supelco

Clonazolam-D4 solution

100 μg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
R$1,047.00

R$1,047.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
R$1,047.00

About This Item

Empirical Formula (Hill Notation):
C17H8D4ClN5O2
Molecular Weight:
357.79
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

R$1,047.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=C([2H])C([2H])=C([2H])C([2H])=C1C2=NCC3=NN=C(C)N3C4=C2C=C([N+]([O-])=O)C=C4

InChI

1S/C17H12ClN5O2/c1-10-20-21-16-9-19-17(12-4-2-3-5-14(12)18)13-8-11(23(24)25)6-7-15(13)22(10)16/h2-8H,9H2,1H3/i2D,3D,4D,5D

InChI key

XJRGLCAWBRZUFC-QFFDRWTDSA-N

General description

Clonazolam, also known as clonitrazolam, is a highly potent benzodiazepine analog that can cause strong sedation and amnesia at minimal oral doses. Clonazolam is also sold on the designer drug market with cases of abuse noted in Europe and the US. This internal standard is suitable for Clonazolam testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, urine drug testing, pain prescription monitoring or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service