Skip to Content
Merck
All Photos(1)

Key Documents

C-133

Supelco

2C-B-FLY-D4 hydrochloride solution

100 μg/mL in acetonitrile: water (9:1) (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10D4BrNO2 · HCl
CAS Number:
Molecular Weight:
324.63
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland)

concentration

100 μg/mL in acetonitrile: water (9:1) (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

General description

2C-B-FLY is a psychedelic phenethylamine of the 2C amine series which include drugs such as 2C-B, 2C-I, 2C-E and 2C-C. Psychedelic effects of this designer drug usually last from 6-8 hours with up to 12 hours observed for larger doses. This stable labeled internal standard is suitable for quantitation of 2C-B-FLY levels in LC/MS or GC/MS applications including forensic analysis, clinical toxicology, and urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service