Skip to Content
Merck
All Photos(1)

Key Documents

999988P

Avanti

13:0 Diether PC

Avanti Research - A Croda Brand 999988P, powder

Synonym(s):

1,2-di-O-tridecyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$2,483.00

R$2,483.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
R$2,483.00

About This Item

Empirical Formula (Hill Notation):
C34H72NO6P
CAS Number:
Molecular Weight:
621.91
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

R$2,483.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (999988P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 999988P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OCCCCCCCCCCCCC)COCCCCCCCCCCCCC)=O

InChI

1S/C34H72NO6P/c1-6-8-10-12-14-16-18-20-22-24-26-29-38-32-34(33-41-42(36,37)40-31-28-35(3,4)5)39-30-27-25-23-21-19-17-15-13-11-9-7-2/h34H,6-33H2,1-5H3/t34-/m1/s1

InChI key

BEBKMIWGRRQYKZ-UUWRZZSWSA-N

General description

13:0 Diether PC (phosphocholine) is a class of glycerophospholipids that has two tridecane chains attached to the sn-1 and sn-2 positions of the glycerol backbone by ether bonds. It has choline as the alcohol moiety, attached to the phosphate group.[1]

Application

13:0 Diether PC or 1,2-di-O-tridecyl-sn-glycero-3-phosphocholine is suitable for bicelle preparation[2] and as an internal standard for lipid identification in tissue.[3]

Biochem/physiol Actions

13:0 Diether PC (phosphocholine) is useful for bicelles preparation. Bicelles can be integrated into standard crystallization protocols and in contrast to micelles, bicelles maintain the protein in a more native bilayer environment allowing proteins to be captured in a more biologically relevant orientation.

Packaging

5 mL Amber Glass Screw Cap Vial (999988P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Baumeier et al.
Biochimica et biophysica acta, 1851(5), 566-576 (2015-02-04)
Caloric restriction and intermittent fasting are known to improve glucose homeostasis and insulin resistance in several species including humans. The aim of this study was to unravel potential mechanisms by which these interventions improve insulin sensitivity and protect from type
Hugo F Azurmendi et al.
Carbohydrate research, 337(10), 905-915 (2002-05-15)
The conformations of two synthetic trisaccharides of blood group A and B (alpha-L-Fucp-(1-->2)-[alpha-D-GalpNAc-(1-->3)]-alpha-D-Galp and alpha-L-Fucp-(1-->2)-[alpha-D-Galp-(1-->3)]-alpha-D-Galp, respectively) and of a type A tetrasaccharide alditol, Fucp-(1-->2)-[alpha-D-GalpNAc-(1-->3)]-beta-D-Galp-(1-->3)-GalNAc-ol, were studied by NMR measurements of one-bond C-H residual dipolar couplings in partially oriented liquid crystal
John M Dean et al.
Protein & cell, 9(2), 196-206 (2017-05-20)
Ether lipids, such as plasmalogens, are peroxisome-derived glycerophospholipids in which the hydrocarbon chain at the sn-1 position of the glycerol backbone is attached by an ether bond, as opposed to an ester bond in the more common diacyl phospholipids. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service