Skip to Content
Merck
All Photos(1)

Key Documents

900109E

Avanti

PGE2-EA-d4

Avanti Research - A Croda Brand

Synonym(s):

Prostaglandin E2 Ethanolamide(d4)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H33D4NO5
CAS Number:
Molecular Weight:
399.56
UNSPSC Code:
12352100
NACRES:
NA.12

Assay

>99% (TLC)

form

ethanol solution

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

pkg of (pkg of 1 X 100 ug (900109E-100UG))

concentration

100 μg/mL in ethanol

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@H]1CC([C@H](C/C=C\CCCC(NC([2H])(C(O)([2H])[2H])[2H])=O)[C@H]1/C=C/[C@@H](O)CCCCC)=O

Packaging

2 mL Amber Glass Sealed Ampule (900109E-100UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

WGK

""

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service