Skip to Content
Merck
All Photos(1)

Key Documents

880620P

Avanti

18:0 PEG750 PE

Avanti Research - A Croda Brand 880620P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-750] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
R$969.00
200 MG
R$2,066.00

R$969.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
R$969.00
200 MG
R$2,066.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

R$969.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 200 mg (880620P-200mg)
pkg of 1 × 25 mg (880620P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880620P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+]

InChI key

QAVSYMFTABOITA-CQZUBJGASA-N

General description

18:0 PEG750 PE is a polyethylene glycol (PEG) conjugated phospholipid. The molecular weight and exact mass are averages based on the polydispersity of PEG.

Application

18:0 PEG750 PE might be used to PEGylate lipoplexes and to study the feasibility of its entrapment into a solid matrix system for prolonged delivery of siRNA in vaginal mucus.[1]

Biochem/physiol Actions

18:0 PEG750 PE is useful as a micelle-forming component.[2]

Packaging

20 mL Clear Glass Screw Cap Vial (880620P-200mg)
5 mL Clear Glass Sealed Ampule (880620P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tania Furst et al.
Journal of controlled release : official journal of the Controlled Release Society, 236, 68-78 (2016-06-23)
Topical vaginal sustained delivery of siRNA presents a significant challenge due to the short residence time of formulations. Therefore, a drug delivery system capable to adhere to the vaginal mucosa is desirable, as it could allow a prolonged delivery and
Rupa R Sawant et al.
International journal of pharmaceutics, 374(1-2), 114-118 (2009-05-19)
Cell-penetrating peptide (TATp) was attached to the distal tips of polyethyleneglycol (PEG) moieties of polyethyleneglycol-phosphatidylethanolamine (PEG-PE) micelles loaded with paclitaxel (PCT). The TATp-modified micelles demonstrated an increased interaction with cancer cells compared to non-modified micelles resulting in a significant increase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service