Skip to Content
Merck
All Photos(1)

Documents

880152P

Avanti

DSG-PEG 2000

Avanti Research - A Croda Brand

Synonym(s):

Distearoyl-rac-glycerol-PEG2K

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C130H258O50
CAS Number:
Molecular Weight:
2621.41
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 g (880152P-1g)
pkg of 5 × 1 g (880152P-5G)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

advanced drug delivery

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(CCCCCCCCCCCCCCCCC)OCC(OC(CCCCCCCCCCCCCCCCC)=O)COCCOCCOC

Looking for similar products? Visit Product Comparison Guide

Packaging

20 mL Clear Glass Screw Cap Vial (880152P-1g)
30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (880152P-5g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stabilized plasmid--lipid particles containing PEG-diacylglycerols exhibit extended circulation lifetimes and tumor selective gene expression
Ambegia E, et al.
Biochimica et Biophysica Acta - Biomembranes, 1669(2), 155-163 (2005)
Adam Zmysłowski et al.
Food chemistry, 329, 127132-127132 (2020-06-07)
It has been proven that at increased temperature, sterols can undergo various chemical reactions e.g., oxidation, dehydrogenation, dehydration and polymerisation. The objectives of this study are to prove the existence of dimers and to quantitatively analyse the dimers (3β,3'β-disteryl ethers).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service