Skip to Content
Merck
All Photos(1)

Documents

880123X

Avanti

DSPE-PEG(5000) Folate

Avanti Research - A Croda Brand 880123X

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[folate(polyethylene glycol)-5000] (ammonium salt):methanol (9:1)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (880123X-500ug)

manufacturer/tradename

Avanti Research - A Croda Brand 880123X

concentration

0.5 mg/mL (880123X-500ug)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC([(OCH2CH2)113]NC(CC[C@H](NC(C1=CC=C(NCC(C=N2)=NC3=C2N=C(N)NC3=O)C=C1)=O)C([O-])=O)=O)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+].[NH4+]

General description

DSPE-PEG(5000) Folate(1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[folate(polyethylene glycol)-5000]) is incorporated into liposomes as a targeting ligand for folate receptors, which acts as cellular surface markers for various cancers and myeloid leukemias. Folate targeted drug delivery is potentially used for the treatment and imaging of several cancers and inflammatory diseases. Folate conjugates are small molecules having high binding affinity for folate receptors (FR) on cell surface. Therefore, they have an ability to deliver a variety of molecular complexes to pathologic cells without affecting normal tissues.

Application

DSPE-PEG(5000) Folate may be used:
  • in preparation of folate functionalized nanoparticles (NPs)
  • in synthesis of BTPEPBI-containing nanoparticles
  • in preparation of paclitaxel loaded folate modified lipid-shell and polymer-core nanoparticles (FLPNPs)

Packaging

5 mL Clear Glass Sealed Ampule (880123X-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aggregation-induced red-NIR emission organic nanoparticles as effective and photostable fluorescent probes for bioimaging
Zhao Q, et al.
Journal of Materials Chemistry, 22(30), 15128-15135 (2012)
Linhua Zhang et al.
International journal of nanomedicine, 10, 2101-2114 (2015-04-07)
The purpose of this study was to develop a novel lipid-polymer hybrid drug carrier comprised of folate (FA) modified lipid-shell and polymer-core nanoparticles (FLPNPs) for sustained, controlled, and targeted delivery of paclitaxel (PTX). The core-shell NPs consist of 1) a
Folate-modified lipid--polymer hybrid nanoparticles for targeted paclitaxel delivery
Zhang L, et al.
International journal of nanomedicine, 10, 2101-2101 (2015)
Junlong Geng et al.
Small (Weinheim an der Bergstrasse, Germany), 8(23), 3655-3663 (2012-08-16)
Folate functionalized nanoparticles (NPs) that contain fluorogens with aggregation-induced emission (AIE) characteristics are fabricated to show bright far-red/near-infrared fluorescence, a large two-photon absorption cross section and low cytotoxicity, which are internalized into MCF-7 cancer cells mainly through caveolae-mediated endocytosis. One-photon
Andrew R Hilgenbrink et al.
Journal of pharmaceutical sciences, 94(10), 2135-2146 (2005-09-02)
Folate targeted drug delivery has emerged as an alternative therapy for the treatment and imaging of many cancers and inflammatory diseases. Due to its small molecular size and high binding affinity for cell surface folate receptors (FR), folate conjugates have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service