Skip to Content
Merck
All Photos(1)

Key Documents

870817P

Avanti

N-palmitoylglycine

Avanti Research - A Croda Brand

Synonym(s):

Glycine, N-(1-oxohexadecyl)-

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$248.00

R$248.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
R$248.00

About This Item

Empirical Formula (Hill Notation):
C18H35NO3
CAS Number:
Molecular Weight:
313.48
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

R$248.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (870817P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

neutral glycerides
neutral lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(CCCCCCCCCCCCCCC)NCC(O)=O

InChI

1S/C18H35NO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-17(20)19-16-18(21)22/h2-16H2,1H3,(H,19,20)(H,21,22)

InChI key

KVTFEOAKFFQCCX-UHFFFAOYSA-N

General description

N-palmitoylglycine belongs to the family of acyl amides, which are endogenous lipids with a potential to modulate pain and inflammation. Structurally, N-palmitoylglycine (PalGly) comprises 16-carbon saturated fatty acid attached to glycine through amide linkage. It is a structural analog of phospholipid-derived N-acyl ethanolamine.[1] N-palmitoylglycine, a saturated lipid is endogenously produced at high levels in rat skin and spinal cord.[1][2]

Application

N-palmitoylglycine has been used as an intact polar lipid (IPL) standard for determination of δ 15N values of nitrogen containing headgroups of IPLs using gas chromatography/combustion/isotope-ratio mass spectrometry.[3]

Biochem/physiol Actions

N-palmitoylglycine stimulates calcium influx and nitric oxide (NO) production through calcium-sensitive nitric-oxide synthase in sensory neurons.[1] It acts as a lipid activator of G-protein-coupled receptor GPR132.[4]

Packaging

5 mL Amber Glass Screw Cap Vial (870817P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisabeth Svensson et al.
Rapid communications in mass spectrometry : RCM, 29(23), 2263-2271 (2015-11-03)
Compound-specific isotope analysis (CSIA) of nitrogen in amino acids has proven a valuable tool in many fields (e.g. ecology). Several intact polar lipids (IPLs) also contain nitrogen, and their nitrogen isotope ratios have the potential to elucidate food-web interactions or
Cyrine Ezzili et al.
Bioorganic & medicinal chemistry letters, 20(20), 5959-5968 (2010-09-08)
Key studies leading to the discovery and definition of the role of endogenous fatty acid amide signaling molecules are summarized.
Neta Rimmerman et al.
Molecular pharmacology, 74(1), 213-224 (2008-04-22)
N-arachidonoyl glycine is an endogenous arachidonoyl amide that activates the orphan G protein-coupled receptor (GPCR) GPR18 in a pertussis toxin (PTX)-sensitive manner and produces antinociceptive and antiinflammatory effects. It is produced by direct conjugation of arachidonic acid to glycine and
Shalini Chaturvedi et al.
Prostaglandins & other lipid mediators, 81(3-4), 136-149 (2006-11-07)
Oleamide (cis-9-octadecenamide) is a member of an emerging class of lipid-signaling molecules, the primary fatty acid amides. A growing body of evidence indicates that oleamide mediates fundamental neurochemical processes including sleep, thermoregulation, and nociception. Nevertheless, the mechanism for oleamide biosynthesis
James R Foster et al.
Pharmacology research & perspectives, 7(6), e00542-e00542 (2019-11-27)
The G-protein-coupled receptor GPR132, also known as G2A, is activated by 9-hydroxyoctadecadienoic acid (9-HODE) and other oxidized fatty acids. Other suggested GPR132 agonists including lysophosphatidylcholine (LPC) have not been readily reproduced. Here, we identify N-acylamides in particular N-acylglycines, as lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service