Skip to Content
Merck
All Photos(1)

Key Documents

870741P

Avanti

18:1 ether Coenzyme A

Avanti Research - A Croda Brand 870741P, powder

Synonym(s):

(9Z-octadecenyl) Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,293.00

R$1,293.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$1,293.00

About This Item

Empirical Formula (Hill Notation):
C39H79N10O16P3S
CAS Number:
Molecular Weight:
1069.09
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

R$1,293.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870741P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870741P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSCCCCCCCC/C=C\CCCCCCCC)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C39H70N7O16P3S.3H3N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-23-66-24-22-41-30(47)20-21-42-37(50)34(49)39(2,3)26-59-65(56,57)62-64(54,55)58-25-29-33(61-63(51,52)53)32(48)38(60-29)46-28-45-31-35(40)43-27-44-36(31)46;;;/h11-12,27-29,32-34,38,48-49H,4

InChI key

NKJGEJWYUPZQQY-ROTIMMBTSA-N

General description

18:1 ether Coenzyme A or (9Z-octadecenyl) Coenzyme A, is a thioether analog of (9Z)-octadecenoic acid. It belongs to a group of long-chain acyl-CoA esters. Long-chain acyl-CoA esters are involved in regulation of various processes such as fatty acid synthesis, enzyme systems, vesicle trafficking, ion channels and ion pumps.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (870741P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Elholm et al.
Journal of lipid research, 41(4), 538-545 (2000-04-01)
A detailed analysis of the subcellular distribution of acyl-CoA esters in rat liver revealed that significant amounts of long-chain acyl-CoA esters are present in highly purified nuclei. No contamination of microsomal or mitochondrial marker enzymes was detectable in the nuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service