Skip to Content
Merck
All Photos(1)

Key Documents

870722P

Avanti

22:0 Coenzyme A

Avanti Research - A Croda Brand 870722P, powder

Synonym(s):

behenoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,746.00
10 MG
R$2,617.00

R$1,746.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$1,746.00
10 MG
R$2,617.00

About This Item

Empirical Formula (Hill Notation):
C43H87N10O17P3S
CAS Number:
Molecular Weight:
1141.19
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

R$1,746.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (870722P-10mg)
pkg of 1 × 5 mg (870722P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870722P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCCCCCCCCCCCCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C43H78N7O17P3S.3H3N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-34(52)71-27-26-45-33(51)24-25-46-41(55)38(54)43(2,3)29-64-70(61,62)67-69(59,60)63-28-32-37(66-68(56,57)58)36(53)42(65-32)50-31-49-35-39(44)47-30-48-40(35)50;;;/h30-32,36-38,42,53-54H,4-29H2,1-3H3,(H,45,51)(H,46,55)(H,59,60)(H,61,62)(H2,44,47,48)(H2,56,57,58);3*1H3/t32-,36?,37+,38+,42-;;;/m1.../s1

InChI key

DSSMNGMIRQGXTR-CNAHFMJYSA-N

General description

22:0 Coenzyme A, also known as behenoyl Coenzyme A, is a coenzyme A derivative of behenic acid.

Application

22:0 Coenzyme A has been used as a substrate in fatty acyl-CoA reductase (AmFAR1) enzyme assay[1] and ceramide synthase assay.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (870722P-10mg)
5 mL Amber Glass Screw Cap Vial (870722P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prapapan Teerawanichpan et al.
Insect biochemistry and molecular biology, 40(9), 641-649 (2010-06-15)
Honey bees (Apis mellifera) are social insects which have remarkable complexity in communication pheromones. These chemical signals comprise a mixture of hydrocarbons, wax esters, fatty acids, aldehydes and alcohols. In this study, we detected several long chain aliphatic alcohols ranging
Evgeny V Berdyshev et al.
The Journal of biological chemistry, 284(9), 5467-5477 (2009-01-03)
Novel immunomodulatory molecule FTY720 is a synthetic analog of myriocin, but unlike myriocin FTY720 does not inhibit serine palmitoyltransferase. Although many of the effects of FTY720 are ascribed to its phosphorylation and subsequent sphingosine 1-phosphate (S1P)-like action through S1P(1,3-5) receptors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service