Skip to Content
Merck
All Photos(1)

Key Documents

870621P

Avanti

18:0-PhoDAG

Avanti Research - A Croda Brand, powder

Synonym(s):

1-stearoyl-2-[(E)-4-(4-((4-butylphenyl)diazenyl)phenyl)butanoyl]-sn-glycerol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$3,642.00
25 MG
R$13,780.00

R$3,642.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$3,642.00
25 MG
R$13,780.00

About This Item

Empirical Formula (Hill Notation):
C41H64N2O5
CAS Number:
Molecular Weight:
664.96
UNSPSC Code:
12352211
NACRES:
NA.25

R$3,642.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

1 ea of 25 mg (870621P-25mg)
1 ea of 5 mg (870621P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

photoswitchable lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(OC(CCCC1=CC=C(/N=N/C2=CC=C(CCCC)C=C2)C=C1)=O)COC(CCCCCCCCCCCCCCCCC)=O

InChI

1S/C41H64N2O5/c1-3-5-7-8-9-10-11-12-13-14-15-16-17-18-19-23-40(45)47-34-39(33-44)48-41(46)24-20-22-36-27-31-38(32-28-36)43-42-37-29-25-35(26-30-37)21-6-4-2/h25-32,39,44H,3-24,33-34H2,1-2H3/b43-42+/t39-/m0/s1

InChI key

YOEPDENSWVOMRQ-VGDFDPFWSA-N

General description

PhoDAG is a light-sensitive, photo-switchable diacylglycerol (DAG). It is inactive in the dark and gets activated by ultraviolet-A (UV-A) light. PhoDAG can be deactivated by blue light.[1]

Biochem/physiol Actions

PhoDAG plays an important role in studying the pathophysiological role of diacylglycerol (DAG)-sensitive transient receptor potential (TRP) channels, which are involved in maintaining the homeostasis of the body.[1]

Packaging

5 mL Amber Glass Screw Cap Vial

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PhoDAGs enable optical control of diacylglycerol-sensitive transient receptor potential channels
Leinders-Zufall T, et al.
Cell Chemical Biology, 25(2), 215-223 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service