Skip to Content
Merck
All Photos(1)

Documents

860684P

Avanti

16:1 SM (d18:1/16:1(9Z))

Avanti Research - A Croda Brand

Synonym(s):

N-palmitoleoyl-D-erythro-sphingosylphosphorylcholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H77N2O6P
CAS Number:
Molecular Weight:
701.01
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860684P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCC/C=C\CCCCCC)=O)COP([O-])(OCC[N+](C)(C)C)=O

General description

16:1 SM (d18:1/16:1(9Z)), also known as N-palmitoleoyl-D-erythro-sphingosylphosphorylcholine, is a sphingomyelin containing palmitoleic acid (16:1) ((9Z)-hexadec-9-enoic acid).

Application

16:1 SM (d18:1/16:1(9Z)) or N-palmitoleoyl-D-erythro-sphingosylphosphorylcholine has been used as a standard in organellar lipidome analysis by Raman spectrometry.

Packaging

5 mL Clear Glass Sealed Ampule (860684P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian Lita et al.
Analytical chemistry, 91(17), 11380-11387 (2019-08-06)
Detailed studies of lipids in biological systems, including their role in cellular structure, metabolism, and disease development, comprise an increasingly prominent discipline called lipidomics. However, the conventional lipidomics tools, such as mass spectrometry, cannot investigate lipidomes until they are extracted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service