Skip to Content
Merck
All Photos(1)

Documents

860560C

Avanti

18:1-12:0 Biotin PS

Avanti Research - A Croda Brand 860560C

Synonym(s):

1-oleoyl-2-(12-biotinyl(aminododecanoyl))-sn-glycero-3-phospho-L-serine (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H86N5O12PS
CAS Number:
Molecular Weight:
964.24
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (860560C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860560C

concentration

1 mg/mL (860560C-1mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCCCCCCNC(CCCC[C@]1([H])SC[C@@]2([H])[C@]1([H])NC(N2)=O)=O)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

InChI

1S/C46H83N4O12PS.H3N/c1-2-3-4-5-6-7-8-9-10-11-12-14-17-20-23-30-42(52)59-33-37(34-60-63(57,58)61-35-38(47)45(54)55)62-43(53)31-24-21-18-15-13-16-19-22-27-32-48-41(51)29-26-25-28-40-44-39(36-64-40)49-46(56)50-44;/h9-10,37-40,44H,2-8,11-36,47H2,1H3,(H,48,51)(H,54,55)(H,57,58)(H2,49,50,56);1H3/b10-9-;/t37-,38+,39+,40+,44+;/m1./s1

InChI key

FUCJSUOHIUUCPS-FNICJJOFSA-N

General description

18:1-12:0 Biotin PS is a biotinylated lipid.

Application

18:1-12:0 Biotin PS may be used in lipid blot analysis to study Niemann-Pick C2 (NPC2)-lipid interactions.

Packaging

5 mL Clear Glass Sealed Ampule (860560C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intracellular cholesterol trafficking is dependent upon NPC2 interaction with lysobisphosphatidic acid
McCauliff LA, et al.
eLife, 8 (2019)
Peter A Leventis et al.
Annual review of biophysics, 39, 407-427 (2010-03-03)
Phosphatidylserine (PS) is the most abundant negatively charged phospholipid in eukaryotic membranes. PS directs the binding of proteins that bear C2 or gamma-carboxyglutamic domains and contributes to the electrostatic association of polycationic ligands with cellular membranes. Rather than being evenly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service