Skip to Content
Merck
All Photos(1)

Documents

860481P

Avanti

N-12:0-1-deoxysphinganine

Avanti Research - A Croda Brand 860481P, powder

Synonym(s):

N-dodecanoyl-1-deoxysphinganine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H61NO2
CAS Number:
Molecular Weight:
467.81
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860481P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860481P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Amber Glass Screw Cap Vial (860481P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tania C B Santos et al.
Langmuir : the ACS journal of surfaces and colloids, 36(21), 6007-6016 (2020-05-06)
Compared to the canonical sphingoid backbone of sphingolipids (SLs), atypical long-chain bases (LCBs) lack C1-OH (1-deoxy-LCBs) or C1-CH2OH (1-deoxymethyl-LCBs). In addition, when unsaturated, they present a cis-double bond instead of the canonical  Δ4-5 trans-double bond. These atypical LCBs are directly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service