Skip to Content
Merck
All Photos(2)

Key Documents

860373P

Avanti

18:0 PE-d70

Avanti Research - A Croda Brand 860373P, powder

Synonym(s):

1,2-distearoyl-d70-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$9,726.00

R$9,726.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
R$9,726.00

About This Item

Empirical Formula (Hill Notation):
C41H12NO8PD70
CAS Number:
Molecular Weight:
818.50
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

R$9,726.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860373P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860373P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H

General description

1,2-distearoyl-d70-sn-glycero-3-phosphoethanolamine (18:0 PE-d70) is a deuterated phosphoethanolamine (PE) in which 70 protons of distearoyl are replaced.
Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Application

1,2-distearoyl-d70-sn-glycero-3-phosphoethanolamine (18:0 PE-d70) has been used in the preparation of 1,2-distearoyl-sn-glycero-3-phosphoethanolamine (DSPE) bilayers to determine the pressure dependence of DSPE flip-flop.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860373P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy C Anglin et al.
Biochemistry, 48(43), 10220-10234 (2009-09-15)
In order to better characterize the dependence of lipid flip-flop rate and thermodynamics on the nature of the lipid headgroup, we have studied the kinetics of flip-flop for single-lipid and mixed-lipid bilayers consisting of 1,2-distearoyl-sn-glycero-3-phosphocholine (DSPC) and 1,2-distearoyl-sn-glycero-3-phosphoethanolamine (DSPE) as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service