Skip to Content
Merck
All Photos(2)

Key Documents

860351P

Avanti

16:0 PC-d4

Avanti Research - A Croda Brand, powder

Synonym(s):

1,2-Dipalmitoyl-sn-glycero-3-phosphocholine-1,1,2,2-d4

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$8,037.00

R$8,037.00


Estimated to ship onApril 28, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
R$8,037.00

About This Item

Empirical Formula (Hill Notation):
C40H76NO8PD4
CAS Number:
Molecular Weight:
738.06
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

R$8,037.00


Estimated to ship onApril 28, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860351P-10MG)

manufacturer/tradename

Avanti Research - A Croda Brand

technique(s)

mass spectrometry (MS): suitable

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC([2H])([2H])C([2H])([2H])[N+](C)(C)C)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C40H80NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-39(42)46-36-38(37-48-50(44,45)47-35-34-41(3,4)5)49-40(43)33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h38H,6-37H2,1-5H3/t38-/m1/s1/i34D2,35D2

InChI key

KILNVBDSWZSGLL-QKXUHBRISA-N

Application

16:0 PC-d4 has been used in the preparation of multilamellar lipid dispersions for nuclear magnetic resonance (NMR) studies.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860351P-10MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of diacylglycerols on conformation of phosphatidylcholine headgroups in phosphatidylcholine/phosphatidylserine bilayers
Goldberg E M, et al.
Biophysical Journal, 69(3), 965-973 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service