Skip to Content
Merck
All Photos(2)

Documents

858142P

Avanti

16:0 Lyso PS

1-palmitoyl-2-hydroxy-sn-glycero-3-phospho-L-serine (sodium salt), powder

Synonym(s):

1-hexadecanoyl-sn-glycero-3-phospho-L-serine (sodium salt); PS(16:0/0:0); 110723

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H43NPO9Na
CAS Number:
Molecular Weight:
519.54
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

99% (LPS; may contain up to 15% of the 2-LPS isomer, TLC)

form

powder

packaging

pkg of 1 × 1 mg (858142P-1mg)
pkg of 1 × 5 mg (858142P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 858142P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)([H])COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C22H44NO9P.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-21(25)30-16-19(24)17-31-33(28,29)32-18-20(23)22(26)27;/h19-20,24H,2-18,23H2,1H3,(H,26,27)(H,28,29);/q;+1/p-1/t19-,20+;/m1./s1

InChI key

MNVYLWFGOUZBHG-FDOHDBATSA-M

Application

16:0 Lyso PS has been used to prevent the lack of de novo synthesis of phosphatidylserine (PS) in Golgi apparatus.

Packaging

5 mL Amber Glass Screw Cap Vial (858142P-1mg)
5 mL Amber Glass Screw Cap Vial (858142P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miki Eto et al.
International journal of molecular sciences, 13(12), 16267-16280 (2012-12-05)
Cellular membranes contain glycerophospholipids, which have important structural and functional roles in cells. Glycerophospholipids are first formed in the de novo pathway (Kennedy pathway) and are matured in the remodeling pathway (Lands' cycle). Recently, lysophospholipid acyltransferases functioning in Lands' cycle
Suriakarthiga Ganesan et al.
Traffic (Copenhagen, Denmark), 20(3), 226-245 (2018-12-21)
Diacylglycerol (DAG) is a key signaling lipid and intermediate in lipid metabolism. Our knowledge of DAG distribution and dynamics in cell membranes is limited. Using live-cell fluorescence microscopy we investigated the localization of yeast cytosolic-facing pools of DAG in response
Jung Yong Eum et al.
Journal of chromatography. A, 460849-460849 (2020-01-14)
Aging refers to the intracellular accumulation of reactive oxygen species that damages proteins, DNA, and lipids. As alterations in lipid metabolism may trigger metabolic disorders and the onset of metabolic diseases, changes in lipid profiles can be closely related to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service