Skip to Content
Merck
All Photos(2)

Key Documents

857391P

Avanti

W140

(S)-3-amino-4-(3-hexylphenylamino)-4-oxobutylphosphonic acid (TFA salt), powder

Synonym(s):

S1P1 Control Molecule

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$2,483.00

R$2,483.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
R$2,483.00

About This Item

Empirical Formula (Hill Notation):
C18H28F3N2O6P
CAS Number:
Molecular Weight:
456.39
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

R$2,483.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (857391P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857391P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

General description

W140 or (S)-3-amino-4-(3-hexylphenylamino)-4-oxobutylphosphonic acid is an inactive enantiomer of W146.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (857391P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingjing Liu et al.
Journal of biochemical and pharmacological research, 1(4), 197-203 (2014-11-11)
Sphingosine-1-phosphate (S1P), a serum-borne bioactive lipid, regulates various physiological functions. We observed that the S1P receptor subtype 1 (S1P1), a high affinity G-protein coupled receptor of S1P, is the major S1P receptor expressed in the Kit+/Sca-1+/Lin- (KSL) hematopoietic stem progenitor
Shreya Endapally et al.
Cell, 176(5), 1040-1053 (2019-02-05)
Sphingomyelin and cholesterol are essential lipids that are enriched in plasma membranes of animal cells, where they interact to regulate membrane properties and many intracellular signaling processes. Despite intense study, the interaction between these lipids in membranes is not well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service