Skip to Content
Merck
All Photos(2)

Key Documents

857128P

Avanti

18:0 Lyso PA

Avanti Research - A Croda Brand

Synonym(s):

1-octadecanoyl-sn-glycero-3-phosphate (sodium salt); stearoyl lysophosphatidic acid; PA(18:0/0:0); 18:0 LPA; s-LPA; 110680

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
R$999.00

R$999.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
R$999.00

About This Item

Empirical Formula (Hill Notation):
C21H42O7PNa
CAS Number:
Molecular Weight:
460.52
UNSPSC Code:
51191904
NACRES:
NA.25

R$999.00


Check Cart for Availability

Request a Bulk Order

description

1-stearoyl-2-hydroxy-sn-glycero-3-phosphate (sodium salt)

Assay

>99% (LPA; may contain up to 10% of the 2-LPA isomer, TLC)

form

powder

packaging

pkg of 1 × 25 mg (857128P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(O)=O)([H])COC(CCCCCCCCCCCCCCCCC)=O.[Na+]

InChI key

YJQFVAZHPIBPHN-VEIFNGETSA-M

General description

Lysophosphatidic acid (1- or 2-acyl-lysophosphatidic acid/LPA), a bioactive phospholipid is seen at a low level in plasma (~100nM). It is usually formed during the synthesis of phospholipid of cell membranes. It is produced by various cells like, activated platelets, epithelial cells, leukocytes, neuronal cells and tumor cells.[1] 18:0 Lyso PA has stearic acid at sn-1 and a hydroxyl group at sn-2 position.[2]

Application

18:0 Lyso PA is suitable for use:
  • to determine the extraction recovery of LPA at neutral pH and acidic pH by chromatographic method[3]
  • to use as internal standard to study the role of intestinally derived lysophosphatidic acid in dyslipidemia and atherosclerosis[2]
  • to investigate its effects on clathrin?dependent endocytosis[4]

Biochem/physiol Actions

Lysophosphatidic acid (1- or 2-acyl-lysophosphatidic acid/LPA) modulates cell motility, proliferation, invasion, survival and production of growth factors via G protein-coupled receptor (GPCR).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (857128P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Lysophosphatidic acid triggers apoptosis in HeLa cells through the upregulation of tumor necrosis factor receptor superfamily member 21
Dong Y, et al.
Mediators of Inflammation, 2017 (2017)
Exogenous lysophospholipids with large head groups perturb clathrin-mediated endocytosis
Ailte I, et al.
Traffic, 18(3), 176-191 (2017)
Quantitative determination of lysophosphatidic acid by LC/ESI/MS/MS employing a reversed phase HPLC column
Shan L, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 864(1-2), 22-28 (2008)
Mohamad Navab et al.
Journal of lipid research, 54(12), 3403-3418 (2013-10-03)
We recently reported that levels of unsaturated lysophosphatidic acid (LPA) in the small intestine significantly correlated with the extent of aortic atherosclerosis in LDL receptor-null (LDLR⁻/⁻) mice fed a Western diet (WD). Here we demonstrate that WD increases unsaturated (but
Olaposi I Omotuyi et al.
Scientific reports, 5, 13343-13343 (2015-08-14)
Lysophosphatidic acid (LPA) receptor 1 (LPA1) is a member of the G protein-coupled receptors mediating the biological response to LPA species. Lack of detailed mechanism underlying LPA/LPA1 interaction has hampered the development of specific antagonists. Here, novel N-terminal Lys39 has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service