Skip to Content
Merck
All Photos(1)

Key Documents

850560P

Avanti

MAG Detergent Kit

powder

Synonym(s):

Detergent Kit for Lipid Mesophase Crystallization

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$19,943.00

R$19,943.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
R$19,943.00

About This Item

UNSPSC Code:
12161902
NACRES:
NA.28

R$19,943.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

The MAG Detergent Kit is designed to allow for the convenient evaluation of 8 unique MAG detergent reagents for their ability to influence lipid mesophase crystallization. This method has variously been referred to as the lipidic cubic phase or in meso method. The method has been shown to be quite versatile in that it has been used to solve X-ray crystallographic structures of prokaryotic and eukaryotic proteins, proteins that are monomeric, homo- and hetero-multimeric, chromophore-containing and chromophore-free, and alpha-helical and beta-barrel proteins.

Packaging

5ML PTFE Vial with Screw Cap for each component (850560P-1EA))(8)
Package contains 200MG of each: 7.7 MAG, 7.8 MAG, 7.9 MAG, 8.6 MAG, 8.7 MAG, 8.8 MAG, 8.9 MAG, 9.7 MAG

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service