Skip to Content
Merck
All Photos(2)

Key Documents

850398P

Avanti

22:1 (Cis) PC

1,2-dierucoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1,2-di-(13Z-docosenoyl)-sn-glycero-3-phosphocholine; PC(22:1(13Z)/22:1(13Z)); DEPC

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
R$550.00

R$550.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
R$550.00

About This Item

Empirical Formula (Hill Notation):
C52H100NO8P
CAS Number:
Molecular Weight:
898.33
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

R$550.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (850398P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850398P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCC/C=C\CCCCCCCC)=O)=O

InChI

1S/C52H100NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38-40-42-44-51(54)58-48-50(49-60-62(56,57)59-47-46-53(3,4)5)61-52(55)45-43-41-39-37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h20-23,50H,6-19,24-49H2,1-5H3/b22-20+,23-21+/t50-/m1/s1

InChI key

SDEURMLKLAEUAY-QVOKQPLLSA-N

General description

The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

22:1 (Cis) PC has been used as in the preparation of model membrane mimicking T-cell (LM3).[1] It is suitable for use to study about lipid dynamics and also to investigate the effect of cholesterol on it.[2]

Biochem/physiol Actions

22:1 (Cis) PC/1,2-dierucoyl-sn-glycero-3-phosphocholine (DEPC)[3] is a longer lipid.[4] It is 7 Å thicker, when compared to 1,2-dioleoyl-sn-glycero-3-phosphocholine (DOPC).[1]

Packaging

5 mL Clear Glass Sealed Ampule (850398P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HIV fusion peptide penetrates, disorders, and softens T-cell membrane mimics
Tristram NS, et al.
Journal of Molecular Biology, 402(1), 139-153 (2010)
How phospholipid-cholesterol interactions modulate lipid lateral diffusion, as revealed by fluorescence correlation spectroscopy
Kahya N and Schwille P
Journal of Fluorescence, 16(5), 671-678 (2006)
Dynamic assessment of bilayer thickness by varying phospholipid and hydraphile synthetic channel chain lengths
Weber ME, et al.
Journal of the American Chemical Society, 127(2), 636-642 (2005)
Gareth R Tibbs et al.
The Journal of pharmacology and experimental therapeutics, 345(3), 363-373 (2013-04-04)
Chronic pain after peripheral nerve injury is associated with afferent hyperexcitability and upregulation of hyperpolarization-activated, cyclic nucleotide-regulated (HCN)-mediated IH pacemaker currents in sensory neurons. HCN channels thus constitute an attractive target for treating chronic pain. HCN channels are ubiquitously expressed;
Dependence of M13 major coat protein oligomerization and lateral segregation on bilayer composition
Fernandes F, et al.
Biophysical Journal, 85(4), 2430-2441 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service