790326C
Avanti
Fluorescent Transfection Reagent
Avanti Research™ - A Croda Brand 790326C
Synonym(s):
Fluorescent Transfection Reagent, Fluorescent Transfection Reagent, contains DOTAP:DOPE (1:1, w/w) + 0.1 wt.% NBD-DOPE
About This Item
form
liquid
packaging
pkg of 1 × 1 mL (790326C-10mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 790326C
concentration
10 mg/mL (790326C-10mg)
application(s)
advanced drug delivery
lipid type
cationic lipids
transfection
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Packaging
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service