Skip to Content
Merck
All Photos(1)

Key Documents

790106P

Avanti

16:0 PE-DTPA

Avanti Research - A Croda Brand 790106P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-diethylenetriaminepentaacetic acid (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$655.00

R$655.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$655.00

About This Item

Empirical Formula (Hill Notation):
C51H110N9O17P
CAS Number:
Molecular Weight:
1152.44
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

R$655.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (790106P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790106P

shipped in

dry ice

storage temp.

−20°C

General description

This chelate can be valuable for preparing MRI contrast agents.

Packaging

5 mL Amber Glass Screw Cap Vial (790106P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lan Wu et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 152, 108-115 (2020-05-22)
Pulmonary formulations have been attracting much attention because of their direct effects on respiratory diseases, but also their non-invasive administration for the treatment of systemic diseases. When developing such formulations, they are typically first investigated in mice. As there are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service