Skip to Content
Merck
All Photos(1)

Documents

700125P

Avanti

DMHCA

Avanti Research - A Croda Brand 700125P, powder

Synonym(s):

N,N-dimethyl-3β-hydroxycholenamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H43NO2
CAS Number:
Molecular Weight:
401.63
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700125P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700125P

shipped in

dry ice

storage temp.

−20°C

General description

N,N-dimethyl-3β-hydroxy-cholenamide (DMHCA) is a potent steroidal liver X receptor (LXR) ligand in vitro and in vivo. It is used to treat atherosclerosis.
N,N-dimethyl-3β-hydroxycholenamide (DHMCA) is a synthetic ligand for liver X receptor (LXR).

Application

DMHCA has been used as LXR activator to treat retinal endothelial cells in order to activate the sirtuin 1- liver X receptors (SIRT1-LXR) pathway.
It may be used as 3β-hydroxysterol-Δ24-reductase (DHCR24) inhibitor.

Biochem/physiol Actions

N,N-dimethyl-3β-hydroxycholenamide (DHMCA) halts cholesterol biosynthesis by inhibiting 3β-hydroxysterol-Δ24-reductase (DHCR24) enzyme. It may serve as a therapeutic agent for treating atherosclerosis, age-related macular degeneration and diabetic retinopathy.
DMHCA (N,N-dimethyl-3β-hydroxycholenamide) is a synthetic oxysterol and is an agonist for liver X receptors (LXR). It promotes cholesterol transport, expression of adenosine triphosphate (ATP) binding cassette transporters. DMHCA suppresses the proinflammatory cytokines stimulation, induced by oxysterol.

Packaging

5 mL Amber Glass Screw Cap Vial (700125P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic LXR agonist attenuates plaque formation in apoE-/-mice without inducing liver steatosis and hypertriglyceridemia
Kratzer A, et al.
Journal of Lipid Research, 50(2), 312-326 (2009)
Gene-selective modulation by a synthetic oxysterol ligand of the liver X receptor.
Quinet E M, et al.
Journal of Lipid Research, 45(10), 1929-1942 (2004)
The mechanism of diabetic retinopathy pathogenesis unifying key lipid regulators, sirtuin 1 and liver X receptor
Hammer SS, et al.
EBioMedicine, 22, 181-190 (2017)
Liver X receptor agonist inhibits proliferation of ovarian carcinoma cells stimulated by oxidized low density lipoprotein
Scoles DR, et al.
Gynecologic Oncology, 116(1), 109-116 (2010)
Thomas Pfeifer et al.
Current pharmaceutical biotechnology, 12(2), 285-292 (2010-12-31)
The liver X receptors (LXRs) are key regulators of genes involved in cholesterol homeostasis. Natural ligands and activators of LXRs are oxysterols. Numerous steroidal and non-steroidal synthetic LXR ligands are under development as potential drugs for individuals suffering from lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service