Skip to Content
Merck
All Photos(1)

Key Documents

700124P

Avanti

Chol-C10-PC

Avanti Research - A Croda Brand 700124P, powder

Synonym(s):

10-cholesteryloxy decylphosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$878.00

R$878.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$878.00

About This Item

Empirical Formula (Hill Notation):
C42H78NO5P
CAS Number:
Molecular Weight:
708.05
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

R$878.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700124P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700124P

shipped in

dry ice

storage temp.

−20°C

General description

Cholesteryl phosphocholine (CholPC) structure has hydrophobic region and a large polar head group.[1] Chol-C10-PC (10-cholesteryloxy decylphosphocholine) has decylphosphocholine attached to cholesterol.

Biochem/physiol Actions

The presence of phosphocholine in the head group of cholesteryl phosphocholine (CholPC) enables shielding of the lipid complex from interaction with water.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (700124P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Max Lönnfors et al.
Langmuir : the ACS journal of surfaces and colloids, 29(7), 2319-2329 (2013-01-30)
We prepared cholesteryl phosphocholine (CholPC) by chemical synthesis and studied its interactions with small (ceramide and cholesterol) and large headgroup (sphingomyelin (SM) and phosphatidylcholine) colipids in bilayer membranes. We established that CholPC could form bilayers (giant uni- and multilamellar vesicles
Kacee H Sims et al.
Biochimica et biophysica acta, 1841(8), 1038-1048 (2014-06-24)
Lipid research is challenging owing to the complexity and diversity of the lipidome. Here we review a set of experimental tools developed for the seasoned lipid researcher, as well as, those who are new to the field of lipid research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service