600310AVT
Avanti
60 mL Boston Round
Avanti Research™ - A Croda Brand
Synonym(s):
60 mL amber narrow mouth bottle with PTFE lined screw cap closure
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
24122000
NACRES:
NA.25
Recommended Products
manufacturer/tradename
Avanti Research™ - A Croda Brand
storage temp.
room temp
Packaging
pkg of 1 x 60 mL bottle (600310-1EA)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service