Skip to Content
Merck
All Photos(1)

Documents

ATE443546277

AstaTech, Inc.

2-AMINO-1-FURAN-2-YL-ETHANOL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
127.14
MDL number:

mp

85-87 C

Iupac Name

2-amino-1-(2-furyl)ethanol

Inchi Code

1S/C6H9NO2/c7-4-5(8)6-2-1-3-9-6/h1-3,5,8H,4,7H2

InChI key

JWQAFPHYLSGNSK-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=54490

Melting Point

85-87 C

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=71472


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service