APO455833967
Apollo Scientific Ltd
5-Fluoro-2-methylphenylacetic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
120-123
Iupac Name
(5-fluoro-2-methylphenyl)acetic acid
Inchi Code
1S/C9H9FO2/c1-6-2-3-8(10)4-7(6)5-9(11)12/h2-4H,5H2,1H3,(H,11,12)
InChI key
BPWHUOQJJZVQLO-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Purity
97%
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid-Powder
Product Link
https://store.apolloscientific.co.uk/product/5-fluoro-2-methylphenylacetic-acid
Synonym
5-Fluoro-o-tolylacetic acid, 2-(Carboxymethyl)-4-fluorotoluene
Boiling point
120-123
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/5-fluoro-2-methylphenylacetic-acid
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service