Skip to Content
Merck
All Photos(1)

Documents

APO455828458

Apollo Scientific Ltd

Methyl 3-amino-4-chlorobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
185.61
MDL number:

bp

82-83

Iupac Name

methyl 3-amino-4-chlorobenzoate

Inchi Code

1S/C8H8ClNO2/c1-12-8(11)5-2-3-6(9)7(10)4-5/h2-4H,10H2,1H3

InChI key

LOCJPOYKBUUVKU-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Powder

Product Link

https://store.apolloscientific.co.uk/product/methyl-3-amino-4-chlorobenzoate

Boiling point

82-83

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/methyl-3-amino-4-chlorobenzoate


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service