Z743787
ART® Non-Filtered Low Retention Tip
ART 20, size 20 μL, hinged rack, sterile
Synonym(s):
aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
sterility
sterile
feature
ART 20
hinged rack
packaging
pkg of 5X960 ea
manufacturer/tradename
Molecular BioProducts 3521-05-HR
technique(s)
PCR: suitable
size
20 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service